Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRDE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100410 Supplier Invitrogen™ Supplier No. PA5100410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C14orf102.

NRDE2 gene ontology (GO) annotations include RNA interference; chromatin silencing by small RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRDE2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NRDE2
Gene Accession No. Q80XC6, Q9H7Z3
Gene Alias 6720454P05Rik; C14orf102; Nrde2; nrde-2 necessary for RNA interference, domain containing; NRDE-2, necessary for RNA interference, domain containing; Nuclear exosome regulator NRDE2; Protein NRDE2 homolog; UPF0614 protein C14orf102; UPF0614 protein C14orf102 homolog
Gene Symbols NRDE2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NRDE2(Accession Q9H7Z3), corresponding to amino acid residues E568-D618.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217827, 55051
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.