Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NREP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568426
Description
NREP Polyclonal Antibody for Western Blot, IHC (P)
P311, also known as C5orf13 (chromosome 5 open reading frame 13), D4S114, PTZ17 or PRO1873, is a 68 amino acid cytoplasmic protein involved in cellular differentiation, neural function and axonal regeneration. Found in the granular layer of the cerebellum, P311 is expressed at lower levels in hippocampus, olfactory bulb, kidney, liver and heart and when expressed ectopically, P311 augments giloma motility. P311 is enriched in mice within the superficial cortical layers and striatum at E20 and the germinal zones at E17. Known to interact with Filamin 1, P311 regulates retinoic-acid lipid-droplet biogenesis, induces myofibroblast ameboid migration and the differentiation of fibroblasts into myofibroblasts. Ser-59 phosphorylation decreases P311 stability; the gene encoding P311 maps to human chromosome 5q22.1.
Specifications
NREP | |
Polyclonal | |
Unconjugated | |
NREP | |
AI325076; C5orf13; D0H4S114; D4S114; Harp; high-altitude regulated protein; neuronal protein 3.1; neuronal regeneration related protein; neuronal regeneration related protein homolog; neuronal regeneration-related protein; Nrep; P311; PRO1873; Protein p311; PTZ17; SEZ17 | |
Rabbit | |
affinity chromatography | |
RUO | |
9315 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q16612 | |
NREP | |
Synthesized peptide derived from human NREP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction