Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Nrf1 Antibody (NRF1/2608), Janelia Fluor™ 646, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308751JF646
Description
Nrf1 Monoclonal antibody specifically detects Nrf1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Nrf1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Alpha palindromic-binding protein, ALPHA-PAL, EWG, NRF-1, nuclear respiratory factor 1 | |
| Recombinant full-length human Nrf1 protein (Uniprot: Q16656) | |
| 100 μg | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| 4899 | |
| Store at 4°C in the dark. | |
| IgG1 κ | 
| Immunohistochemistry (Paraffin), Protein Array | |
| NRF1/2608 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction