Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nrf1 Antibody (NRF1/2609), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308773AF594
Description
Nrf1 Monoclonal antibody specifically detects Nrf1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Nrf1 | |
Monoclonal | |
Alexa Fluor 594 | |
Alpha palindromic-binding protein, ALPHA-PAL, EWG, NRF-1, nuclear respiratory factor 1 | |
Recombinant full-length human Nrf1 protein (Uniprot: Q16656) | |
100 μg | |
Endocrinology, Neurodegeneration, Neuroscience | |
4899 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
NRF1/2609 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction