Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583082 Supplier Invitrogen™ Supplier No. PA583082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATVATLADAS ELPTTVTVAQ VNYSAVADGE VEQNWATLQG GEMTIQTTQA SEATQAVASL AEAAVAASQE MQQGATVTMA LNSEAAAHAV ATLAEATLQG GGQIVLSGET AAAVGALTGV QDANGLFMAD RAGRKWILTD Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 96%.

This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for 'nuclear factor (erythroid-derived 2)-like 1' which has an official symbol of NFE2L1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRF1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NRF1
Gene Accession No. Q16656
Gene Alias Alpha palindromic-binding protein; alpha-pal; C87038; D6Ertd415e; EWG; LOW QUALITY PROTEIN: nuclear respiratory factor 1; Nfe2l1_retired; Not really finished protein; nrf; Nrf1; NRF-1; nrf1 protein; nrl; nuclear respiratory factor 1; nuclear respiratory factor-1; SC:BZ3C13.1; SC:bZ3C13.2; transcription factor; ube2h
Gene Symbols NRF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NRF1. Recombinant protein control fragment (Product #RP-89320).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.