Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRF2 Mouse anti-Human, Clone: 6H11.1, MilliporeSigma™

Catalog No. MABC1556MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1556MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1556MI Supplier MilliporeSigma Supplier No. MABC1556
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect NRF2 using this mouse monoclonal Anti-NRF2 Antibody, clone 6H11.1. It has been used in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRF2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H11.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols NFE2L2,NRF2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Nuclear factor erythroid 2-related factor 2 (NRF2).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NRF2
Test Specificity Clone 6H11.1 detects NRF2 in HepG2 cells. It targets an epitope within 15 amino acids in the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.