Learn More
NRF2 NFE2L2 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The NRF2, NFE2L2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NRF2, NFE2L2. This antibody recognizes human, mouse, rat antigen. The NRF2, NFE2L2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
Specifications
| Antigen | NRF2 NFE2L2 |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Conjugate | CoraLite 594 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 4780 |
| Target Species | Human, Rat, Mouse |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.