Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nrf2, p Ser40 Antibody (2E6), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326402100UL
Description
Nrf2 Monoclonal antibody specifically detects Nrf2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
Nrf2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
2E6 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
HEBP1, NF-E2-related factor 2, NFE2-related factor 2, NRF2 erythroid derived 2, like 2, nuclear factor (erythroid-derived 2)-like 2, nuclear factor erythroid 2-related factor 2, nuclear factor erythroid-derived 2-like 2 | |
A synthesized peptide derived from Human Nrf2 [p Ser40] [UniProt Q16236] | |
100 μL | |
Phospho Specific, Transcription Factors and Regulators | |
4780 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction