Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nrf2 Rat anti-Rat, Mouse, Clone: 103, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABE1799
Description
Anti-Nrf2, clone 103 is a highly specific rat monoclonal antibody that targets Nuclear factor erythroid 2-related factor 2 and has been tested in Immunohistochemistry and Western Blotting.
Specifications
Nrf2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Nfe2l2,Nrf-2,Nrf2 | |
GST-tagged recombinant fragment corresponding to 179 amino acids from internal region of murine Nuclear factor erythroid 2-related factor 2. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_035032 | |
Rat, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
103 | |
Unconjugated | |
Rat | |
Protein G Purified | |
RUO | |
Primary | |
Clone 103 detects Nrf2 in Keap-1 knockout cells. It targets an epitope within 179 amino acids in the internal region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction