Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRF3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305800MFV500
Description
NRF3 Polyclonal antibody specifically detects NRF3 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
NRF3 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
NF-E2-related factor 3, NFE2-related factor 3, Nrf3, NRF3nuclear factor erythroid 2-related factor 3, nuclear factor (erythroid-derived 2)-like 3, Nuclear factor, erythroid derived 2, like 3, nuclear factor-erythroid 2 p45-related factor 3, nuclear factor-erythroid 2-related factor 3 | |
Produced in rabbits immunized with E. coli-derived Human NRF3 fragment. | |
0.1 mL | |
Stem Cell Markers | |
9603 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction