Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NRG1 beta-1 ELISA Kit

Catalog No. ehnrg1
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

The Human NRG1beta-1 (Hu NRG1) ELISA quantitates Hu NRG1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NRG1. Principle of the method The Human NRG1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Neuregulin 1 (NRG1) was originally identified as a 44-kD glycoprotein that interacts with the NEU/ERBB2 receptor tyrosine kinase to increase its phosphorylation on tyrosine residues. It is known that an extraordinary variety of different isoforms are produced from the NRG1 gene by alternative splicing. These isoforms include heregulins (HRGs), glial growth factors (GGFs) and sensory and motor neuron-derived factor (SMDF). They are tissue-specifically expressed and differ significantly in their structure. The HRG isoforms all contain immunoglobulin (Ig) and epidermal growth factor-like (EGF-like) domains. GGF and GGF2 isoforms contain a kringle-like sequence plus Ig and EGF-like domains; and the SMDF isoform shares only the EGF-like domain with other isoforms. The receptors for all NRG1 isoforms are the ERBB family of tyrosine kinase transmembrane receptors. Through interaction with ERBB receptors, NRG1 isoforms induce the growth and differentiation of epithelial, neuronal, glial, and other types of cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02297
Assay Range 82.3 to 20,000 pg/mL
Assay Sensitivity 50 pg/mL
Calibration Range 50 pg/mL to 20000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family activation of transmembrane receptor protein tyrosine kinase activity; cardiac conduction system development; cardiac muscle cell differentiation; cardiac muscle cell myoblast differentiation; cell communication; cell migration; cell morphogenesis; cell proliferation; cellular protein complex disassembly; embryo development; endocardial cell differentiation; epidermal growth factor receptor signaling pathway; ERBB signaling pathway; Fc-epsilon receptor signaling pathway; fibroblast growth factor receptor signaling pathway; glial cell fate commitment; innate immune response; locomotory behavior; mammary gland development; MAPK cascade; negative regulation of cardiac muscle cell apoptotic process; negative regulation of extrinsic apoptotic signaling pathway in absence of ligand; negative regulation of protein catabolic process; negative regulation of secretion; negative regulation of transcription, DNA-templated; nervous system development; neural crest cell development; neuron fate commitment; neurotransmitter receptor metabolic process; neurotrophin TRK receptor signaling pathway; peripheral nervous system development; phosphatidylinositol-mediated signaling; positive regulation of cardiac muscle cell proliferation; positive regulation of cell adhesion; positive regulation of cell growth; positive regulation of protein kinase B signaling; positive regulation of protein tyrosine kinase activity; positive regulation of Ras protein signal transduction; positive regulation of striated muscle cell differentiation; protein tyrosine kinase activator activity; regulation of protein heterodimerization activity; regulation of protein homodimerization activity; synapse assembly; transcription cofactor activity; transmembrane receptor protein tyrosine kinase activator activity; transmembrane receptor protein tyrosine kinase signaling pathway; ventricular cardiac muscle cell differentiation; ventricular trabecula myocardium morphogenesis; wound healing
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3084
Gene Symbol ARIA,GGF,GGF2,HGL,HRG,HRG1,HRGA,MST131,MSTP131,NDF-IT2,SMDF,NRG1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF-IT2, SMDF, NRG1
Target NRG1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.