Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRG1 Mouse anti-Human, Unconjugated, Clone: NRG1/2752, Abnova™

Catalog No. 89337715 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-715 Supplier Abnova Corporation Supplier No. MAB21222
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

NRG1 Monoclonal antibody specifically detects NRG1 in Human samples. It is validated for WB, ELISA.

Specifications

Antigen NRG1
Applications Western Blot, ELISA
Classification Monoclonal
Clone NRG1/2752
Conjugate Unconjugated
Description neuregulin 1
Gene NRG1
Gene Alias ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, NDF, SMDF
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 21-242 of human NRG1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3084
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.