Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565487 Supplier Invitrogen™ Supplier No. PA565487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVIRRETETTF SGSHSCSPSH HCSTATPTSS HRHESHTWSL ERSESLTSDS QSGIMLSSVG TSKCNSPACV EARARRAAAY NLEERRRATA PPYHDSVDSL Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

Neuregulin 2 is a novel member of the neuregulin family of growth and differentiation factors. Through interaction with the Erbb family of receptors, NRG2 induces the growth and differentiation of epithelial, neuronal, glial, and other types of cells. The gene consists of 12 exons and the genomic structure is similar to that of neuregulin 1, another member of the neuregulin family of ligands. NRG1 and NRG2 mediate distinct biological processes by acting at different sites in tissues and eliciting different biological responses in cells. The gene is located close to the region for demyelinating Charcot-Marie-Tooth disease locus, but is not responsible for this disease. Alternative transcripts encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRG2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NRG2
Gene Accession No. O14511
Gene Alias divergent of neuregulin-1; Don1; DON-1; HRG2; neural- and thymus-derived activator for ErbB kinases; neuregulin 2; Neuregulin-2; NRG2; NRG-2; NTAK; NTAK alpha1; NTAK_alpha2a; pro-neuregulin-2, membrane-bound isoform; Pro-NRG2
Gene Symbols NRG2
Host Species Rabbit
Immunogen Recombinant Human NRG2. Recombinant protein control fragment (Product #RP-104841).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9542
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.