Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103013 Supplier Invitrogen™ Supplier No. PA5103013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NRG3.

This gene is a member of the neuregulin gene family. This gene family encodes ligands for the transmembrane tyrosine kinase receptors ERBB3 and ERBB4 - members of the epidermal growth factor receptor family. Ligand binding activates intracellular signaling cascades and the induction of cellular responses including proliferation, migration, differentiation, and survival or apoptosis. This gene encodes neuregulin 3. NRG3 has been shown to activate the tyrosine phosphorylation of its cognate receptor, ERBB4, and is thought to influence neuroblast proliferation, migration and differentiation by signalling through ERBB4. NRG3 also promotes mammary differentiation during embryogenesis. Linkage studies have implicated this gene as a susceptibility locus for schizophrenia and schizoaffective disorder. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described but their biological validity has not been verified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRG3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NRG3
Gene Accession No. O35181, P56975
Gene Alias HGNC:7999; HRG3; neuregulin 3; neuregulin 3 variant 1; neuregulin 3 variant 10; neuregulin 3 variant 11; neuregulin 3 variant 12; neuregulin 3 variant 13; neuregulin 3 variant 14; neuregulin 3 variant 3; neuregulin 3 variant 4; neuregulin 3 variant 5; neuregulin 3 variant 6; neuregulin 3 variant 7; neuregulin 3 variant 8; neuregulin 3 variant 9; Neuregulin-3; neuregulin-3-like polypeptide; NRG3; NRG-3; Pro-neuregulin-3, membrane-bound isoform; pro-NRG3; RGD1559678; scaramanga; ska
Gene Symbols NRG3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NRG3(Accession P56975), corresponding to amino acid residues T689-K720.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10718, 18183
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.