Learn More
Invitrogen™ NRG3 Recombinant Rabbit Monoclonal Antibody (JE49-84)

Description
NRG3 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | NRG3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JE49-84 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | NRG3 |
| Gene Accession No. | O35181, P56975 |
| Gene Alias | HGNC:7999; HRG3; neuregulin 3; neuregulin 3 variant 1; neuregulin 3 variant 10; neuregulin 3 variant 11; neuregulin 3 variant 12; neuregulin 3 variant 13; neuregulin 3 variant 14; neuregulin 3 variant 3; neuregulin 3 variant 4; neuregulin 3 variant 5; neuregulin 3 variant 6; neuregulin 3 variant 7; neuregulin 3 variant 8; neuregulin 3 variant 9; Neuregulin-3; neuregulin-3-like polypeptide; NRG3; NRG-3; Pro-neuregulin-3, membrane-bound isoform; pro-NRG3; RGD1559678; scaramanga; ska |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.