Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRG4 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299949F
Description
NRG4 Polyclonal specifically detects NRG4 in Human samples. It is validated for ELISA.Specifications
NRG4 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
145957 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4 | |
Produced in rabbits immunized with purified, recombinant NRG4 (Q8WWG1; Pro2-Phe62). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction