Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRGN Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550991
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550991 200 μL
FEMA550992 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550991 Supplier Invitrogen™ Supplier No. MA550991
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRGN
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene NRGN
Gene Accession No. P60761, Q04940, Q92686
Gene Alias 0710001B06Rik; AI838505; B-50 immunoreactive C-kinase substrate; BICKS; calmodulin-binding protein; hng; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); ng; NG/RC3; NRGN; p17; postsynaptic spine 1; protein kinase C substrate; protein kinase C substrate 7.5 kDa protein; protein kinase C substrate, RC3; Pss1; R75334; RC3; similar to neurogranin
Gene Symbols NRGN
Host Species Mouse
Immunogen Recombinant Human NRGN protein, Met1-Ala67 (Accession #Q92686).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100738830, 4900, 64011, 64356
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.