Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRH2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987915
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7915 Supplier Bioss Supplier No. BS7672R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a secreted peptide hormone and member of the gonadotropin-releasing hormone (GnRH) family of proteins. The encoded protein regulates reproductive function by stimulating the production and release of the gonadotropins follicle-stimulating hormone (FSH) and luteinizing hormone (LH). The encoded protein may inhibit endometrial, ovarian, prostate, and breast cancer cell proliferation. Alternative splicing results in multiple transcript variants.

Specifications

Antigen NRH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNRH2
Gene Accession No. O43555
Gene Alias GnRH II; GNRH2; GnRH-associated peptide 2; GnRH-associated peptide II; GnRH-II; Gonadoliberin II; Gonadoliberin-2; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 2; Gonadotropin-releasing hormone II; LH-RH II; LH-RHII; Luliberin II; Luteinizing hormone-releasing hormone II; Progonadoliberin II; Progonadoliberin-2
Gene Symbols GNRH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from rat NRH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2797
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.