Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRIF3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen NRIF3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159187
SDP
View Documents
Novus Biologicals
NBP159187
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

NRIF3 Polyclonal specifically detects NRIF3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NRIF3
Polyclonal
Rabbit
Apoptosis
Q13352
23421
Synthetic peptides corresponding to ITGB3BP(integrin beta 3 binding protein (beta3-endonexin)) The peptide sequence was selected from the N terminal of ITGB3BP (NP_055103). Peptide sequence TGTCQMSLFASPTSSEEQKHRNGLSNEKRKKLNHPSLTESKESTTKDNDE.
Primary
Western Blot
Unconjugated
RUO
Human
beta-3-endonexin, CENP-R, centromere protein R, HSU37139, integrin beta 3 binding protein (beta3-endonexin), Integrin beta-3-binding protein, NRIF3CENPRbeta 3 endonexin, Nuclear receptor-interacting factor 3, TAP20
ITGB3BP
IgG
20 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.