Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRIF3 Antibody (OTI4H1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245461
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
NRIF3 Monoclonal specifically detects NRIF3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NRIF3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:2000 | |
beta-3-endonexin, CENP-R, centromere protein R, HSU37139, integrin beta 3 binding protein (beta3-endonexin), Integrin beta-3-binding protein, NRIF3CENPRbeta 3 endonexin, Nuclear receptor-interacting factor 3, TAP20 | |
Mouse | |
20 kDa | |
0.1 mL | |
Apoptosis | |
23421 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4H1 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
ITGB3BP | |
Full length human recombinant protein of human ITGB3BP (NP_055103) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction