Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRIP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572624
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2624 10 μL
50-157-2623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2624 Supplier Bethyl Laboratories, Inc Supplier No. IHC00735T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey, Gorilla, Chimpanzee, Small-eared galago and White-tufted-ear marmoset

NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up regulates their transcriptional activity, thereby functioning as a nuclear receptor coactivator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer thus indicating a therapeutic intervention. Northern Blot analysis detected a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.

Specifications

Antigen NRIP
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DCAF6
Gene Accession No. Q58WW2
Gene Alias ARCAP, DCAF6, IQWD1, MSTP055, DDB1- and CUL4-associated factor 6, Androgen receptor complex-associated protein, Nuclear receptor interaction protein, IQ motif and WD repeat-containing protein 1
Gene Symbols DCAF6
Host Species Rabbit
Immunogen Between 600 and 650
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55827
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.