Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589510 Supplier Invitrogen™ Supplier No. PA589510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, HT-29, 293T, Mouse brain, Rat brain; Cellular Location: Nucleus Immunogen sequence: QDLIHGQESR RKTSRTEIPA LLVNCKCQDQ LLRVAVDTGT QYNRISAGCL SRLGLEKRVL KASAGDLAPG PPTQVEQLEL QLGQETVVCS AQVVDAESPE FCLGLQTLLS LKCCIDLEHG VLRLKAPFSE LPFLPLYQEP GQ.

NRIP2 down-regulates transcriptional activation by nuclear receptors such as NR1F2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRIP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene NRIP2
Gene Accession No. Q9BQI9, Q9JHR9
Gene Alias AW491344; DKFZP761G1913; fj77f06; neuronal interacting factor 1; neuronal-interacting factor X 1; Nix1; Nrip2; nuclear receptor interacting protein 2; nuclear receptor interacting protein 2-like; nuclear receptor-interacting protein 2; si:dkey-245f7.1; similar to nuclear receptor interacting protein 2; wu:fj77f06
Gene Symbols NRIP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 140-281 of human NRIP2 (NP_1136621).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60345, 689619, 83714
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.