Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NRIP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566212
Description
Immunogen sequence: ALVDTGCLYNL ISLACVDRLG LKEHVKSHKH EGEKLSLPRH LKVVGQIEHL VITLGSLRLD CPAAVVDDNE KNLSLGLQT Highest antigen sequence identity to the following orthologs - mouse 91%, rat 90%.
NRIP3 is a protein coding gene.
Specifications
NRIP3 | |
Polyclonal | |
Unconjugated | |
Nrip3 | |
A330103B05Rik; C11orf14; D7H11orf14; ICRFP703B1614Q5.2; ICRFP703N2430Q5.2; NRIP3; nuclear receptor interacting protein 3; nuclear receptor-interacting protein 3; NY-SAR-105; Sarcoma antigen NY-SAR-105 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56675 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQ35 | |
Nrip3 | |
Recombinant Human NRIP3. Recombinant protein control fragment (Product #RP-105115). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction