Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRIP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 156641AP150UL

Description
NRIP3 is a protein coding gene.Specifications
NRIP3 | |
Polyclonal | |
Unconjugated | |
NRIP3 | |
C11orf14, NRIP3, NY SAR 105, Sarcoma antigen NY SAR 105 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361625, 56675, 78593 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JJR9, Q9NQ35 | |
Nrip3 | |
NRIP3 Fusion Protein Ag8097 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction