Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRK Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298072APC
Description
NRK Polyclonal antibody specifically detects NRK in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
NRK | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
DKFZp686A17109, EC 2.7.11, EC 2.7.11.1, FLJ16788, MGC131849, NESK, Nik related kinase, nik-related protein kinase | |
Produced in rabbits immunized with E. coli-derived Human NRK fragment. | |
0.1 mL | |
Protein Kinase | |
203447 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction