Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100912 Supplier Invitrogen™ Supplier No. PA5100912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NRK.

The mouse ortholog of this gene encodes a protein kinase required for JNK activation. The encoded protein may be involved in the induction of actin polymerization in late embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NRK
Gene Accession No. Q7Z2Y5, Q9R0G8
Gene Alias AA407323; AA409510; AI225895; AV168563; AW538185; Nck-interacting kinase-like embryo specific kinase; Nesk; Nik related kinase; NIK-like embryo-specific kinase; nik-related protein kinase; Nrk
Gene Symbols NRK
Host Species Rabbit
Immunogen A synthesized peptide derived from human NRK(Accession Q7Z2Y5), corresponding to amino acid residues K292-I342.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203447, 27206
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.