Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NRN1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559713
Description
Immunogen sequence: GELETICRSW NDFHACASQV LSGCPEEAAA VWESLQQEAR QAPRPNNLHT LCGAPVHVRE RGTGSETNQ Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 67%.

Specifications
NRN1L | |
Polyclonal | |
Unconjugated | |
NRN1L | |
MRCC2446; neuritin 1 like; neuritin 1-like; neuritin-like protein; NRN1L; UNQ2446; UNQ2446/PRO5725 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123904 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q496H8 | |
NRN1L | |
Recombinant protein corresponding to Human NRN1L. Recombinant protein control fragment (Product #RP-98585). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction