Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRP1 Mouse anti-Human, Clone:5B1E11, Abnova™

Catalog No. 89333099 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-099 Supplier Abnova Corporation Supplier No. MAB16734
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human NRP1.

NRP1 is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor (VEGF; MIM 192240) and semaphorin (see SEMA3A; MIM 603961) family members. NRP1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration, and invasion.[supplied by OMIM]

Specifications

Antigen NRP1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5B1E11
Conjugate Unconjugated
Description neuropilin 1
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NRP1
Gene Alias BDCA4, CD304, DKFZp686A03134, DKFZp781F1414, NP1, NRP, VEGF165R
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acid 45-59 of human NRP1.
Quantity 100 μg
Gene ID (Entrez) 8829
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.