Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRSF Rabbit anti-Human, Mouse, Rat, Clone: 8R8Z4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | NRSF |
---|---|
Clone | 8R8Z4 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NRSF Monoclonal antibody specifically detects NRSF in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
NRSF | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Stem Cell Markers, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5978 | |
IgG | |
Affinity purified |
8R8Z4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human NRSF (Q13127). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title