Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRSN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101707 Supplier Invitrogen™ Supplier No. PA5101707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NRSN1.

May play an important role in neural organelle transport, and in transduction of nerve signals or in nerve growth. May play a role in neurite extension. May play a role in memory consolidation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRSN1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NRSN1
Gene Accession No. P97799, Q8IZ57
Gene Alias neurensin 1; neurensin-1; Neuro-p24; nrsn1; p24; vesicular membrain protein p24; vesicular membrane protein of 24 kDa; Vesicular membrane protein p24; Vmp
Gene Symbols NRSN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NRSN1(Accession Q8IZ57), corresponding to amino acid residues M1-A39.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140767, 22360, 291129
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.