Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ nrv2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532170 Supplier Invitrogen™ Supplier No. PA532170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Drosophila lysate. Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This is the non-catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of Na(+) and K(+) ions across the plasma membrane. The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen nrv2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene nrv2
Gene Accession No. Q24048
Gene Alias 42kDa; CG9261; CG9261-PA; CG9261-PC; CG9261-PD; CG9261-PE; CG9261-PF; CG9261-PG; Dmel\CG9261; Dmel_CG9261; ecta; ectatic; GH13134p; IP16413p; K-ATPase; l(2)k04223; l(2)k13315; l(2)SH0329; l(2)SH2 0329; Na; Na,K-ATPase beta-subunit; Na[+]/K[+] ATPase; Na[+]/K[+]ATPase; Na[+]/K[+]-ATPase; Na+/K+ ATPase; NaK ATPase; nervana; nervana 2; nervana2; NEST:bs13c10; nirvana; nirvana2; Nrv; nrv2; nrv2.2; nrv2-PA; nrv2-PC; nrv2-PD; nrv2-PE; nrv2-PF; nrv2-PG; Protein nervana 2; Sodium/potassium-dependent ATPase subunit beta-2; Sodium/potassium-transporting ATPase subunit beta-2
Gene Symbols nrv2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Drosophila nrv2.1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 33953
Target Species Drosophila
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.