Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRXN1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100219
Description
NRXN1 Polyclonal specifically detects NRXN1 in Human samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA.Specifications
NRXN1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DKFZp313P2036, FLJ35941, KIAA0578Neurexin I-alpha, neurexin 1, neurexin I, Neurexin I-beta, neurexin-1-alpha, neurexin-1-beta, PTHSL2 | |
Goat | |
Affinity Purified | |
RUO | |
9378 | |
Human | |
IgG |
Western Blot, Flow Cytometry, ELISA | |
0.5 mg/mL | |
Western Blot 0.5 - 1 μg/mL, Flow Cytometry 10 μg/mL, Peptide ELISA Detection limit 1:128000 | |
Q9ULB1 | |
NRXN1 | |
Peptide with sequence C-KEKQPSSAKSSNKN corresponding to C-Terminus according to NP_620072.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_004792.1 and NP_620072.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction