Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRXN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598362 Supplier Invitrogen™ Supplier No. PA598362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRXN1 Polyclonal Antibody for Western Blot, ELISA

Neuronal cell surface protein that may be involved in cell recognition and cell adhesion by forming intracellular junctions through binding to neuroligins. May play a role in formation or maintenance of synaptic junctions. May mediate intracellular signaling. May play a role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRXN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nrxn1
Gene Accession No. P58400, Q9ULB1
Gene Alias 1700062G21Rik; 9330127H16Rik; A230068P09Rik; alpha-latrotoxin receptor (calcium-dependent); DKFZp313P2036; FLJ35941; Hs.22998; Kiaa0578; mKIAA0578; neurexin 1; Neurexin 1alpha; neurexin I; neurexin I-alpha; Neurexin I-beta; neurexin-1; Neurexin-1-alpha; neurexin-1-beta; non-processed neurexin I-alpha; Nrxn1; Nrxn1b; PTHSL2; SCZD17
Gene Symbols Nrxn1
Host Species Rabbit
Immunogen Recombinant Human Neurexin-1 protein (31-310AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9378
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.