Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRXN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116301
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116301 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116301 Supplier Invitrogen™ Supplier No. PA5116301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRXN3 Polyclonal Antibody for Western Blot

Neurexins comprise a family of neuronal cell surface proteins, which include neurexin I (NRXN1), neurexin II (NRXN2), neurexin III (NRXN3) and Caspr (neurexin IV). Neurexins I-III are expressed as a and b isoforms. The a isoforms are made of three cassettes, which contain two LNS (Laminin A, Neurexins, Sex hormone-binding)-domains separated by EGF domains, followed by a transmembrane region and a 55 amino acid cytoplasmic C-terminal. The a isoforms bind to neurexophilins at the second LNS site and to the excitatory neurotoxin a-latrotoxin. The b isoforms have only one LNS-domain, bind to neuroligins, and play a role in the formation and remodeling of synapes. Caspr (for Contactin-Associated Protein 1, also designated Paranodin in mouse), contains an extracellular domain similar to the other three neurexins, and binds to the surface glycoprotein Contactin. Caspr and the closely related Caspr2, a mammalian homolog of Drosophila Neurexin IV (Nrx-IV), demarcate distinct subdomains in myelinated axons. Specifically, Caspr exists at the paranodal junctions, while Caspr2 colocalizes with Shaker-like K+ channels in the juxtaparanodal region. Caspr may play a role in the communication of glial cells and neurons during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRXN3
Applications Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NRXN3
Gene Accession No. Q9HDB5, Q9Y4C0
Gene Alias C14orf60; KIAA0743; neurexin 3; Neurexin 3alpha; neurexin III; Neurexin III-alpha; Neurexin III-beta; Neurexin-3; neurexin-3-alpha; Neurexin-3-beta; Neurexin-3-beta, C-terminal fragment; Neurexin-3-beta, soluble form; Nrxn3; NRXN3-CTF
Gene Symbols NRXN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 436-462 amino acids from the C-terminal region of human NRXN3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9369
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.