Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NS 1738
GREENER_CHOICESDP

Catalog No. 299550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-955-0 50 mg
29-951-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-955-0 Supplier R&D Systems™ Supplier No. 2995/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of α7 nAChR; active in vivo

Selective positive allosteric modulator of α7 nicotinic acetylcholine receptors. Exhibits no substantial activity for α4β2, α3β3 and α1-containing receptors. Displays cognitive-enhancing properties in vivo.

Chemical Identifiers

CAS 501684-93-1
Molecular Formula C14H9Cl2F3N2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 365.13
Purity 98%
Molecular Weight (g/mol) 365.133
Chemical Name or Material NS 1738
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.