Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 19504
GREENER_CHOICE

Catalog No. 527610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-761-0 10 mg
52-765-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-761-0 Supplier Tocris Bioscience™ Supplier No. 5276/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KCa1.1 (BK) channel activator

NS 19504 is a selective activator of large-conductance Ca2+-activated potassium channels (BKCa, KCa1.1) (EC50 = 11 μM). Activates BK channels in native smooth muscle cells from guinea pig bladder and reduces spontaneous phasic contractions in bladder strips ex vivo. Displays no effect on contractions induced by a high K+ concentration or at 65 other receptors tested, including L- and N- type Ca2+ channels, Nav, KATP and hERG channels.

Specifications

Chemical Name or Material 5-[(4-Bromophenyl)methyl]-2-thiazolamine
CAS 327062-46-4
Quantity 10 mg
Target Ca2+-Activated Potassium Channel Activators
Molecular Formula C10H9BrN2S
Formula Weight 269.16
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.