Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NS 2028
GREENER_CHOICESDP

Catalog No. 451750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-175-0 50 mg
45-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-175-0 Supplier R&D Systems™ Supplier No. 4517/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent soluble guanylyl cyclase (sGC) inhibitor

Potent soluble guanylyl cyclase (sGC) inhibitor (Ki = 8 nM). Blocks sGC activity in murine cerebellum induced by S-nitroso-glutathione and NMDA (IC50 values are 17 and 20 nM respectively). Inhibits VEGF-induced cGMP accumulation; abolishes VEGF-induced migration in postcapillary venular endothelial cells (CVEC).

Chemical Identifiers

CAS 204326-43-2
Molecular Formula C9H5BrN2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 269.05
Purity >98%
Molecular Weight (g/mol) 269.05
Chemical Name or Material NS 2028
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.