Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 3623
GREENER_CHOICESDP

Catalog No. 446250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-625-0 50 mg
44-6210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-625-0 Supplier Tocris Bioscience™ Supplier No. 4462/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV11.1 (hERG) and KV4.3 channel activator; antiarrhythmic

KV11.1 (hERG) and KV4.3 channel activator. Activates the IKr and Ito currents and displays antiarrhythmic activity. May also act as a partial blocker of KV11.1 channels. Displays selectivity over the key cardiac potassium channels, KV7.1 (KCNQ1) and KV1.5. Suitable for both in vitro and in vivo use.

Chemical Identifiers

CAS 343630-41-1
Molecular Formula C15H10BrF3N6O

Specifications

Quantity 50 mg
Formula Weight 427.18
Purity >99%
Molecular Weight (g/mol) 427.185
Chemical Name or Material NS 3623
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.