Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 3728
GREENER_CHOICE

Catalog No. 697310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-731-0 10 mg
69-735-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-731-0 Supplier Tocris Bioscience™ Supplier No. 6973/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

VRAC blocker

NS 3728 is a volume-regulated anion channel (VRAC) blocker (IC50 = 460 nM). Blocks red blood cell chloride conductance (IC50 = 0.6 μM). NS 3728 also inhibits Ca2+-activated Cl- current (CaCC) by anoctamin (ANO) in an apparently voltage-dependent manner (IC50 against ANO1 = 2.1 and 0.7 μM at -95 and +50 mV, respectively).

Specifications

Chemical Name or Material N-[3,5-bis(Trifluoromethyl)phenyl]-N'-[4-bromo-2-(2 H-tetrazol-5-yl)phenyl]urea
CAS 265646-85-3
Quantity 10 mg
Target Other Chloride Channel Blockers
Molecular Formula C16H9BrF6N6O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.