Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 398
GREENER_CHOICE

Catalog No. 094210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-421-0 10 mg
09-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-421-0 Supplier Tocris Bioscience™ Supplier No. 0942/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cyclooxygenase-2 (COX-2) inhibitor

NS 398 is a selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100 μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.

Specifications

Chemical Name or Material N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide
CAS 123653-11-2
Assay Percent Range >99%
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C13H18N2O5S
Formula Weight Observed MW: 314.36

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.