Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 9283
GREENER_CHOICE

Catalog No. 511210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-121-0 10 mg
51-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-121-0 Supplier Tocris Bioscience™ Supplier No. 5112/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of α4β2 nAChRs

NS 9283 is a positive allosteric modulator of α4β2 receptor; increases the potency of Ach-evoked currents ∼60 fold without affecting the maximum efficacy (HEK293-hα4β2 cells). Reduces the rate of recovery from desensitization and slows the rate of deactivation. Displays a synergistic effect on the Emax when given with NS206 at 3α:2β receptors.

Specifications

Quantity 10 mg
Product Type NS 9283
Purity >99%
Molecular Weight (g/mol) 248.24
Inhibitors NS 9283

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.