Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NS1-BP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563715
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3715 Supplier Bethyl Laboratories, Inc Supplier No. A302879A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Plays a role in cell division and in the dynamic organization of the actin skeleton as a stabilizer of actin filaments by association with F-actin through Kelch repeats. Protects cells from cell death induced by actin destabilization; Protects neurons from dendritic spines and actin filaments damage induced by the actin-destabilizing cytochalasin B when overexpressed. Activates Erk signaling pathway when overexpressed in cultured cell lines (By similarity). May be a component of the cellular splicing machinery with a role in pre-mRNA splicing; may mediate the inhibition of splicing by NS/influenza virus NS1A protein. Functions as modifier of the AHR/Aryl hydrocarbon receptor pathway increasing the concentration of AHR available to activate transcription.

Specifications

Antigen NS1-BP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene IVNS1ABP
Gene Accession No. Q9Y6Y0
Gene Alias aryl hydrocarbon receptor-associated 3, aryl hydrocarbon receptor-associated protein 3, FLARA3, HSPC068, kelch-like family member 39, KIAA0850, KLHL39, NCX downstream gene 1, ND1, NS-1, NS1-binding protein, NS1BP, NS1-BP
Gene Symbols IVNS1ABP
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10625
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.