Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NS5ATP9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987881
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7881 Supplier Bioss Supplier No. BS7627R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCNA-binding protein that acts as a regulator of DNA repair during DNA replication. Following DNA damage, the interaction with PCNA is disrupted, facilitating the interaction between monoubiquitinated PCNA and the translesion DNA synthesis DNA polymerase eta (POLH) at stalled replisomes, facilitating the bypass of replication-fork-blocking lesions. Also acts as a regulator of centrosome number.

Specifications

Antigen NS5ATP9
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PCLAF
Gene Accession No. Q15004
Gene Alias 2810417H13Rik; AA409629; BOS_10802; HCV NS5A-transactivated protein 9; HCV NS5A-transactivated protein 9 homolog; Hepatitis C virus NS5A-transactivated protein 9; KIAA0101; KIAA0101 ortholog; L5; LOC100514810; mKIAA0101; NS5A (hepatitis C virus) transactivated protein 9; Ns5apt9; Ns5atp9; OEATC; OEATC1; OEATC-1; Overexpressed in anaplastic thyroid carcinoma 1; p15(PAF); p15/PAF; p15PAF; PAF; PAF15; PCLAF; PCNA clamp associated factor; PCNA-associated factor; PCNA-associated factor of 15 kDa; PCNA-clamp-associated factor; RIKEN cDNA 2810417H13 gene
Gene Symbols PCLAF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NS5ATP9/PCNA-associated factor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9768
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.