Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NS5B, Mouse anti-Hepatitis C virus (HCV), Clone: 5B-12B7, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16081
Specifications
NS5B | |
Monoclonal | |
Unconjugated | |
In PBS (0.02% sodium azide). | |
Recombinant protein corresponding to HCV NS5B RNA-dependent RNA polymerase. | |
RUO | |
Recognizes HCV NS5B from HCV genotype 1 (epitope mapped to amino acid 139-392 of NS5B). Does not react with genotype 2. | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Western Blot | |
5B-12B7 | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Hepatitis C virus | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction