Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NS5B, Mouse anti-Hepatitis C virus (HCV), Clone: 5B-3B1, Abnova™

Catalog No. 89998890 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-890 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-890 Supplier Abnova Corporation Supplier No. MAB16080
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen NS5B
Applications Western Blot
Classification Monoclonal
Clone 5B-3B1
Conjugate Unconjugated
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (10% glycerol, 0.02% sodium azide).
Host Species Mouse
Immunogen Recombinant protein corresponding to HCV NS5B RNA-dependent RNA polymerase (genotype 1a).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes HCV NS5B (epitope mapped to amino acid 372-382 of NS5B). It recognizes genotype 1 a and also 1b (albeit with lower affinity). It does not recognize genotype 2a.
Target Species Hepatitis C virus
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.