Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hairy Cell Leukemia Monoclonal Antibody (DBA.44)
GREENER_CHOICE

Catalog No. PIMA126274
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126274 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126274 Supplier Invitrogen™ Supplier No. MA126274
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Tonsil.

Hairy Cell Leukemia (HCL) is a rare form of mature B-cell cancer characterized by the presence of abnormal B-cells known as 'hairy cells.' These cells have a distinct genetic profile that plays a significant role in the diagnosis and management of the disease. HCL includes variants such as the hairy cell leukemia variant (HCL-V) and splenic diffuse red pulp lymphoma (SDRPL), which are often detected through specific immunophenotyping and genetic analyses. One critical aspect of these analyses involves the use of flow cytometry to detect lambda light chain restriction among B-cells displaying bright CD11c positivity. Advances in genetic profiling using techniques like clonal analysis with consensus primer polymerase chain reaction (PCR) for heavy chain genes have further refined diagnostic precision, distinguishing HCL from its variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hairy Cell Leukemia
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DBA.44
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.09% sodium azide
Gene NSA2
Gene Accession No. O95478
Gene Alias 5730427N09Rik; CDK105; FLJ94393; Hairy cell leukemia protein 1; HCLG1; HCL-G1; HUSSY29; HUSSY-29; L-name related protein; L-name-related protein 42; LNR42; NSA2; NSA2 ribosome biogenesis homolog; NSA2 ribosome biogenesis homolog (S. cerevisiae); NSA2, ribosome biogenesis homolog; Nsa2p; ribosome biogenesis protein NSA2 homolog; TGF beta-inducible nuclear protein; TGF beta-inducible nuclear protein 1; TGF-beta inducible nuclear protein; TGF-beta-inducible nuclear protein 1; Tinp1; Yr29; YR-29
Gene Symbols NSA2
Host Species Mouse
Immunogen DEAU cell line established from a large cell lymphoma.
Purification Method Purified
Quantity 6 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10412
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.