Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589854 Supplier Invitrogen™ Supplier No. PA589854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPQNEYIELH RKRYGYRLDY HEKKRKKESR EAHERSKKAK KMIGLKAKLY HKQRHAEKIQ MKKTIKMHEK RNTKQKNDEK TPQGAVPAYL LDREGQSRAK VLSNMIKQKR KEKAGKWEVP LPKVRAQGET EVLKVIRTGK RKKKAWKRMV TKVCFVGDGF TRKPPKYERF IRPMGLRFKK AHVTHPELKA TFCLPILGVK KNPSSPLYTT LGVITKGTVI EVNVSELGLV TQGGKVIWGK YAQVTNNPEN DGCINAVLLV; Positive Samples: HeLa, 293T, Mouse heart, Mouse kidney, Mouse skeletal muscle, Rat heart, Rat kidney, Rat skeletal muscle; Cellular Location: Nucleus, nucleolus.

This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. Mice with a mutation that results in reduced expression of the ortholog of this protein are retarded for growth, develop severe tremors by 2 to 3 weeks of age followed by hindlimb paralysis and death by 6 to 10 weeks of age. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NSA2
Gene Accession No. O95478, Q9CR47, Q9QYU7
Gene Alias 5730427N09Rik; CDK105; FLJ94393; Hairy cell leukemia protein 1; HCLG1; HCL-G1; HUSSY29; HUSSY-29; L-name related protein; L-name-related protein 42; LNR42; NSA2; NSA2 ribosome biogenesis homolog; NSA2 ribosome biogenesis homolog (S. cerevisiae); NSA2, ribosome biogenesis homolog; Nsa2p; ribosome biogenesis protein NSA2 homolog; TGF beta-inducible nuclear protein; TGF beta-inducible nuclear protein 1; TGF-beta inducible nuclear protein; TGF-beta-inducible nuclear protein 1; Tinp1; Yr29; YR-29
Gene Symbols NSA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human NSA2 (NP_0557011).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10412, 171456, 59050
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.