Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NSA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100860 Supplier Invitrogen™ Supplier No. PA5100860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TINP1.

This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. Mice with a mutation that results in reduced expression of the ortholog of this protein are retarded for growth, develop severe tremors by 2 to 3 weeks of age followed by hindlimb paralysis and death by 6 to 10 weeks of age. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NSA2
Gene Accession No. O95478, Q9CR47, Q9QYU7
Gene Alias 5730427N09Rik; CDK105; FLJ94393; Hairy cell leukemia protein 1; HCLG1; HCL-G1; HUSSY29; HUSSY-29; L-name related protein; L-name-related protein 42; LNR42; NSA2; NSA2 ribosome biogenesis homolog; NSA2 ribosome biogenesis homolog (S. cerevisiae); NSA2, ribosome biogenesis homolog; Nsa2p; ribosome biogenesis protein NSA2 homolog; TGF beta-inducible nuclear protein; TGF beta-inducible nuclear protein 1; TGF-beta inducible nuclear protein; TGF-beta-inducible nuclear protein 1; Tinp1; Yr29; YR-29
Gene Symbols NSA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NSA2(Accession O95478), corresponding to amino acid residues Q53-S103.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10412, 171456, 59050
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.