Learn More
NSA2/TINP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | NSA2/TINP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9812
![]() |
Bethyl Laboratories, Inc
A305208AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
TINP1 (TGF-beta inducible nuclear protein 1), also known as NSA2 (Nop seven-associated 2), is a highly conserved nucleolar protein essential for large ribosomal subunit biogenesis. NSA2 is broadly expressed in human tissues and cultured cell lines, and located in the nucleolus of the cell. It is thought to be involved in cell proliferation and cell cycle regulation.Specifications
NSA2/TINP1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CDK105, FLJ94393, hairy cell leukemia protein 1, HCLG1, HCL-G1, HUSSY29, HUSSY-29, NSA2 ribosome biogenesis homolog, NSA2 ribosome biogenesis homolog (S. cerevisiae), TGF beta-inducible nuclear protein 1, TGF-beta inducible nuclear protein, TGF-beta-inducible nuclear protein 1, TINP1 | |
Between 150 and 200 | |
Primary | |
4° C | |
NSA2 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O95478, Q9CR47 | |
10412, 59050 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.