Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NSC 23766
GREENER_CHOICE

Catalog No. 216110R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-611-0R 10 mg
21-615-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-611-0R Supplier Tocris Bioscience™ Supplier No. 2161/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of Rac1-GEF interaction; antioncogenic

NSC 23766 is a selective inhibitor of Rac1-GEF interaction. NSC 23766 prevents Rac1 activation by Rac-specific guanine nucleotide exchange factors (GEFs) TrioN and Tiam1 (IC50 ∼ 50 μM) without affecting Cdc42 or RhoA activation. The compound inhibits Rac1-mediated cell functions and is reported to reverse tumor cell phenotypes in prostate cancer cells. NSC 23766 also inhibits mammary carcinoma cell invasion in vitro and inhibits mammary tumor progression and metastasis in a mouse model.

Specifications

Quantity 10 mg
Product Type NSC 23766
Purity >96%
Molecular Weight (g/mol) 530.97
Inhibitors Rho GTPase Inhibitors

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.